Anti-MICALL1 Rabbit Polyclonal Antibody
ABCA_AB220648-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:KIAA1668
- Antigen Symbol:MICALL1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:MIRAB13|MICAL-like protein 1|Molecule interacting with Rab13
- Molecular Weight:93441 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human MICALL1 aa 750 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to MICALL1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MICALL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human