Anti-RUNX3 Rabbit Monoclonal Antibody [clone: EPR20687]
: Abcam
New Product
Rabbit monoclonal [EPR20687] to RUNX3 - ChIP Grade.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RUNX3
Clonality: Monoclonal
Clone: EPR20687
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB224641-1ML
- ABCA_AB224641-100U
- Antigen Synonyms:
- CBFA3|PEBP2A3|Runt-related transcription factor 3|Acute myeloid leukemia 2 protein|Core-binding factor subunit alpha-3|Oncogene AML-2|Polyomavirus enhancer-binding protein 2 alpha C subunit|SL3-3 enhancer factor 1 alpha C subunit|SL3/AKV core-binding factor alpha C subunit|CBF-alpha-3|PEA2-alpha C|PEBP2-alpha C
- CBFA3|PEBP2A3|Runt-related transcription factor 3|Acute myeloid leukemia 2 protein|Core-binding factor subunit alpha-3|Oncogene AML-2|Polyomavirus enhancer-binding protein 2 alpha C subunit|SL3-3 enhancer factor 1 alpha C subunit|SL3/AKV core-binding factor alpha C subunit|CBF-alpha-3|PEA2-alpha C|PEBP2-alpha C
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Host:
- Rabbit
- Rabbit
- ChIP:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 44356 Da
- 44356 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- +4 °C
- +4 °C
- Shipping Temperature:
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
Frequently Bought Together