Gold, nanoparticles, 40 nm
Supplier: Abcam
New Product
Certificates
About this item
40nm Gold Nanoparticles (20 OD) is an aqueous suspension of spherical metallic nanoparticles.
Frequently Bought Together
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
PI MARKER 9.50 200UL
Supplier: 0000014126
Unstained Protein Standard, Broad Range, New England Biolabs
Supplier: New England Biolabs (NEB)
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Potassium iodide
Supplier: G-Biosciences
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
PI MARKER ICE280/ICE3 7.05 200UL
Supplier: 0000014126
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
DNA BINDING BUFFER 100 ML
Supplier: Zymo Research
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript