Anti-p57 Kip2 Rabbit Monoclonal Antibody [clone: EP2718(2)]
ABCA_AB133531-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:KIP2
- Antigen Symbol:p57 Kip2
- Clonality:Monoclonal
- Clone:EP2718(2)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:CDKN1C|Cyclin-dependent kinase inhibitor 1C|Cyclin-dependent kinase inhibitor p57|p57Kip2
- Molecular Weight:32177 Da
- Storage Temperature:Store at –20 °C. Stable for 12 months at –20 °C.
- Shipping Temperature:Shipped at +4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Protein A Affinity purification
- Packaging:Vial
Rabbit monoclonal [EP2718(2)] to p57 Kip2.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: p57 Kip2
Clonality: Monoclonal
Clone: EP2718(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human