Anti-HERPUD1 Rabbit Monoclonal Antibody [clone: EPR9649]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR9649] to HERPUD1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HERPUD1
Clonality: Monoclonal
Clone: EPR9649
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB150424-100U
- ABCA_AB150424-1ML
- Antigen Synonyms:
- KIAA0025|MIF1|Homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domain member 1 protein|Methyl methanesulfonate (MMF)-inducible fragment protein 1
- KIAA0025|MIF1|Homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domain member 1 protein|Methyl methanesulfonate (MMF)-inducible fragment protein 1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Host:
- Rabbit
- Rabbit
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 43720 Da
- 43720 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- +4 °C
- +4 °C
- Shipping Temperature:
- Shipped at +4 °C. Store at –20ºC.
- Shipped at +4 °C. Store at –20ºC.
- Tested Applications:
- Flow Cyt (Intra), ICC/IF
- Flow Cyt (Intra), ICC/IF
Specifications
Frequently Bought Together