Order Entry
United States
ContactUsLinkComponent
Anti-ATP synthase C Rabbit Monoclonal Antibody [clone: EPR13907] (Alexa Fluor® 488)
Anti-ATP synthase C Rabbit Monoclonal Antibody [clone: EPR13907] (Alexa Fluor® 488)
Catalog # ABCA_AB210456-100U
Supplier:  Abcam
New Product
undefined
Anti-ATP synthase C Rabbit Monoclonal Antibody [clone: EPR13907] (Alexa Fluor® 488)
Catalog # ABCA_AB210456-100U
Supplier:  Abcam
Supplier Number:  AB210456-100UL
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    ATP5G1
  • Antigen Symbol:
    ATP synthase C
  • Clonality:
    Monoclonal
  • Clone:
    EPR13907
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 µl
  • Form:
    Liquid
  • Antigen Synonyms:
    ATP5MC1|ATP synthase lipid-binding protein|ATP synthase membrane subunit c locus 1|ATP synthase proteolipid P1|ATP synthase proton-transporting mitochondrial F(0) complex subunit C1|ATPase protein 9|ATPase subunit c
  • Molecular Weight:
    14277 Da
  • Storage Temperature:
    Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle. Store In the Dark.
  • Concentration:
    0.5 mg/ml
  • Shipping Temperature:
    +4 °C
  • Immunogen:
    The exact immunogen used to generate this antibody is proprietary information.
  • Tested Applications:
    ICC/IF
  • Purification:
    Protein A Affinity purification
  • Packaging:
    Vial

Specifications

About this item

Alexa Fluor® 488 Rabbit monoclonal [EPR13907] to ATP synthase C.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ATP synthase C
Clonality: Monoclonal
Clone: EPR13907
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human