Anti-IgA Secretory Component Mouse Monoclonal Antibody [clone: SPM217]
ABCA_AB212330-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Symbol:IgA Secretory Component
- Clonality:Monoclonal
- Clone:SPM217
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Size:100 µg
- Form:Liquid
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Tested Applications:IHC-P
- Purification:Protein A/G Affinity purification
- Packaging:Vial
Mouse monoclonal [SPM217] to IgA Secretory Component - BSA and Azide free.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: IgA Secretory Component
Clonality: Monoclonal
Clone: SPM217
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human