Anti-CD172 gamma Rabbit Monoclonal Antibody [clone: OX117]
Catalog # ABCA_AB245690-200U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CD172g
- Antigen Symbol:CD172 gamma
- Clonality:Monoclonal
- Clone:OX117
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Size:200 µg
- Form:Liquid
- Antigen Synonyms:SIRPB2|SIRPG|Signal-regulatory protein gamma|SIRP-gamma|CD172 antigen-like family member B|Signal-regulatory protein beta-2|SIRP-b2|SIRP-beta-2
- Molecular Weight:42498 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human SIRPG
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit monoclonal [OX117] to CD172 gamma.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD172 gamma
Clonality: Monoclonal
Clone: OX117
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human