Anti-Thiophosphate ester Rabbit Monoclonal Antibody [clone: 51-8]
ABCA_AB92570-50UL
: Abcam
New Product
- Antibody Type:Primary
- Antigen Symbol:Thiophosphate ester
- Clonality:Monoclonal
- Clone:51-8
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Storage Buffer:BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- Storage Temperature:Upon delivery aliquot and store at −20 °C. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Purification:Protein A affinity purification
- Packaging:Vial
Rabbit monoclonal [51-8] to Thiophosphate ester.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Thiophosphate ester
Clonality: Monoclonal
Clone: 51-8
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: