Anti-ErbB2 / HER2 Mouse Monoclonal Antibody [clone: H2Mab-139]
: Abcam
New Product
Mouse monoclonal [H2Mab-139] to ErbB2 / HER2 - BSA and Azide free.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ErbB2 / HER2
Clonality: Monoclonal
Clone: H2Mab-139
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
- Catalog No:
- ABCA_AB264548-100U
- ABCA_AB264548-1MG
- Antigen Symbol:
- ErbB2 / HER2
- ErbB2 / HER2
- Antigen Name:
- CD340
- CD340
- Antigen Synonyms:
- HER2|MLN19|NEU|NGL|ERBB2|Receptor tyrosine-protein kinase erbB-2|Metastatic lymph node gene 19 protein|Proto-oncogene Neu|Proto-oncogene c-ErbB-2|Tyrosine kinase-type cell surface receptor HER2|p185erbB2|MLN 19
- HER2|MLN19|NEU|NGL|ERBB2|Receptor tyrosine-protein kinase erbB-2|Metastatic lymph node gene 19 protein|Proto-oncogene Neu|Proto-oncogene c-ErbB-2|Tyrosine kinase-type cell surface receptor HER2|p185erbB2|MLN 19
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- H2Mab-139
- H2Mab-139
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 137910 Da
- 137910 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Shipped at +4 °C. Store at +4 °C. Do Not Freeze.
- Shipped at +4 °C. Store at +4 °C. Do Not Freeze.
- Shipping Temperature:
- +4 °C
- +4 °C
- Tested Applications:
- IHC-P
- IHC-P
Frequently Bought Together