Order Entry
United States
ContactUsLinkComponent
Anti-S100 alpha 2/S100A2 Rabbit Monoclonal Antibody [clone: EPR5392] (Alexa Fluor® 488)
Anti-S100 alpha 2/S100A2 Rabbit Monoclonal Antibody [clone: EPR5392] (Alexa Fluor® 488)
  ABCA_AB309858-100U
 :  Abcam
New Product
Anti-S100 alpha 2/S100A2 Rabbit Monoclonal Antibody [clone: EPR5392] (Alexa Fluor® 488)
  ABCA_AB309858-100U
 :  Abcam
 :  AB309858-100UL
New Product

 

  • Size:
    100 µl
  • Packaging:
    Vial

 

 

Alexa Fluor® 488 Rabbit monoclonal [EPR5392] to S100 alpha 2/S100A2.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: S100 alpha 2/S100A2
Clonality: Monoclonal
Clone: EPR5392
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: