Anti-SENP2 Mouse Monoclonal Antibody [clone: OTI1H2]
Catalog # ABCA_AB131637-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:KIAA1331
- Antigen Symbol:SENP2
- Clonality:Monoclonal
- Clone:OTI1H2
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Sentrin-specific protease 2|Axam2|SMT3-specific isopeptidase 2|Sentrin/SUMO-specific protease SENP2|Smt3ip2
- Molecular Weight:67855 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human SENP2 aa 100-550. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:Flow Cyt (Intra), ICC/IF, IHC-P
- Purification:Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [OTI1H2] to SENP2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SENP2
Clonality: Monoclonal
Clone: OTI1H2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;African green monkey