Anti-TRAP/CD40L Mouse Monoclonal Antibody [clone: CD40LG/2761]
Catalog # ABCA_AB259254-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CD154
- Antigen Symbol:TRAP/CD40L
- Clonality:Monoclonal
- Clone:CD40LG/2761
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:CD40L|TNFSF5|TRAP|CD40LG|CD40 ligand|CD40-L|T-cell antigen Gp39|TNF-related activation protein|Tumor necrosis factor ligand superfamily member 5
- Molecular Weight:29274 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human CD40LG aa 100 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Protein A/G affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [CD40LG/2761] to TRAP/CD40L - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TRAP/CD40L
Clonality: Monoclonal
Clone: CD40LG/2761
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human