Anti-RPA32/RPA2 Rabbit Polyclonal Antibody
Catalog # ABCA_AB10359-50UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:REPA2
- Antigen Symbol:RPA32/RPA2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse,Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Antigen Synonyms:RPA32|RPA34|RPA2|Replication protein A 32 kDa subunit|RP-A p32|Replication factor A protein 2|Replication protein A 34 kDa subunit|RF-A protein 2|RP-A p34
- Storage Buffer:PBS, tris, citrate
- Molecular Weight:29247 Da
- Storage Temperature:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human RPA2 aa 200 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Immunogen affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to RPA32/RPA2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: RPA32/RPA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human