Anti-Myc tag Mouse Monoclonal Antibody [clone: Myc.A7] (Biotinylated)
Catalog # ABCA_AB173829-50UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Myc Epitope Tag
- Antigen Symbol:Myc tag
- Clonality:Monoclonal
- Clone:Myc.A7
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:c-myc tag
- Storage Temperature:Store at –20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human MYC aa 400 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF
- Purification:Protein G Affinity purification
- Packaging:Vial
Specifications
About this item
Biotin Mouse monoclonal [Myc.A7] to Myc tag.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ADHFE1
Clonality: Monoclonal
Clone: EPR12501
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: