Anti-NF2/Merlin (phospho S518) Rabbit Polyclonal Antibody
ABCA_AB2478-50UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:SCH
- Antigen Symbol:NF2 / Merlin
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Antigen Synonyms:NF2|Merlin|Moesin-ezrin-radixin-like protein|Neurofibromin-2|Schwannomerlin|Schwannomin
- Modification:phospho S518
- Storage Buffer:Potassium phosphate, Sodium chloride
- Molecular Weight:69690 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human NF2 phospho S518 aa 500 - 550 conjugated to Keyhole Limpet Haemocyanin. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Rabbit polyclonal to NF2 / Merlin (phospho S518).
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: NF2 / Merlin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse