Anti-ATP synthase Immunocapture Mouse Monoclonal Antibody [clone: 12F4AD8AF8]
Catalog # ABCA_AB109867-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:ATPI
- Antigen Symbol:ATP synthase Immunocapture
- Clonality:Monoclonal
- Clone:12F4AD8AF8
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:ATPIF1|ATP5IF1|ATP synthase F1 subunit epsilon|Inhibitor of F(1)F(o)-ATPase|IF(1)|IF1
- Molecular Weight:12249 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C. Do Not Freeze.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [12F4AD8AF8] to ATP synthase Immunocapture.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ATP synthase Immunocapture
Clonality: Monoclonal
Clone: 12F4AD8AF8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human