Order Entry
United States
ContactUsLinkComponent
Anti-BTN2A2/BTF2 Mouse Monoclonal Antibody [clone: 6C7D2]
Anti-BTN2A2/BTF2 Mouse Monoclonal Antibody [clone: 6C7D2]
Catalog # ABCA_AB233763-100U
Supplier:  Abcam
New Product
undefined
Anti-BTN2A2/BTF2 Mouse Monoclonal Antibody [clone: 6C7D2]
Catalog # ABCA_AB233763-100U
Supplier:  Abcam
Supplier Number:  AB233763-100UG
New Product

Specifications

  • Size:
    100 µg
  • Packaging:
    Vial

Specifications

About this item

Mouse monoclonal [6C7D2] to BTN2A2/BTF2.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: BTN2A2/BTF2
Clonality: Monoclonal
Clone: 6C7D2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human