Anti-GCAT Rabbit Monoclonal Antibody [clone: EPR13450]
: Abcam
New Product
Rabbit monoclonal [EPR13450] to GCAT - N-terminal.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GCAT
Clonality: Monoclonal
Clone: EPR13450
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB181094-100U
- ABCA_AB181094-1ML
- Antigen Symbol:
- GCAT
- GCAT
- Antigen Name:
- KBL
- KBL
- Antigen Synonyms:
- AKB ligase|Aminoacetone synthase|Glycine acetyltransferase
- AKB ligase|Aminoacetone synthase|Glycine acetyltransferase
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR13450
- EPR13450
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 45285 Da
- 45285 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Concentration:
- 0.576 mg/ml
- 0.576 mg/ml
- Storage Temperature:
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipping Temperature:
- +4 °C
- +4 °C
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P
Frequently Bought Together