Anti-CD59 Mouse Monoclonal Antibody [clone: MEM-43/5]
ABCA_AB9183-100UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:MIC11
- Antigen Symbol:CD59
- Clonality:Monoclonal
- Clone:MEM-43/5
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Western Blot:Yes
- Size:100 µg
- Epitope:The antibody MEM-43/5 reacts with well defined epitope around L33 (see Bodian et al)
- Form:Liquid
- Antigen Synonyms:MIN1|MIN2|MIN3|MSK21|CD59 glycoprotein|1F5 antigen|20 kDa homologous restriction factor|MAC-inhibitory protein|MEM43 antigen|Membrane attack complex inhibition factor|Membrane inhibitor of reactive lysis|Protectin|HRF-20|HRF20|MAC-IP|MACIF|MIRL
- Storage Buffer:PBS
- Molecular Weight:14177 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Cell preparation containing CD59 protein. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Protein A Affinity purification
- Packaging:Vial
Mouse monoclonal [MEM-43/5] to CD59.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MEM-43/5
Conjugation:
Epitope: The antibody MEM-43/5 reacts with well defined epitope around L33 (see Bodian et al)
Host: Mouse
Isotype: IgG2b
Reactivity: Human; Mouse