Anti-Carbonic Anhydrase 1/CA1 Rabbit Monoclonal Antibody [clone: EPR5193]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR5193] to Carbonic Anhydrase 1/CA1.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Carbonic Anhydrase 1/CA1
Clonality: Monoclonal
Clone: EPR5193
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human; Zebrafish
Specifications
- Catalog No:
- ABCA_AB108367-1ML
- ABCA_AB108367-100U
- Antigen Symbol:
- Carbonic Anhydrase 1/CA1
- Carbonic Anhydrase 1/CA1
- Antigen Name:
- Carbonic anhydrase 1
- Carbonic anhydrase 1
- Antigen Synonyms:
- Carbonate dehydratase I|Carbonic anhydrase B|Carbonic anhydrase I|Cyanamide hydratase CA1|CAB|CA-I|CA1
- Carbonate dehydratase I|Carbonic anhydrase B|Carbonic anhydrase I|Cyanamide hydratase CA1|CAB|CA-I|CA1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR5193
- EPR5193
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 28870 Da
- 28870 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Buffer:
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- Storage Temperature:
- Store at –20 °C. Stable for 12 months at –20 °C.
- Store at –20 °C. Stable for 12 months at –20 °C.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Flow Cyt (Intra)
- Flow Cyt (Intra)
Specifications
Frequently Bought Together