Anti-alpha Lactalbumin Goat Polyclonal Antibody HRP (Horseradish Peroxidase)
ABCA_AB112973-1MG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:ALACTA
- Antigen Symbol:alpha Lactalbumin
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:1 mg
- Form:Liquid
- Antigen Synonyms:LALBA|Alpha-lactalbumin|Lactose synthase B protein
- Molecular Weight:16247 Da
- Storage Temperature:Store at +4 °C.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Tested Applications:ICC, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
HRP Goat polyclonal to alpha Lactalbumin.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: alpha Lactalbumin
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine