Anti-Tau Alzheimer’s Disease (phospho S396) Mouse Monoclonal Antibody [clone: GT-38] (Alexa Fluor® 488)
Supplier: Abcam
New Product
Certificates
About this item
Alexa Fluor® 488 Mouse monoclonal [GT-38] to Tau Alzheimer’s Disease - Conformation-Specific.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Tau Alzheimer’s Disease
Clonality: Monoclonal
Clone: GT-38
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB317594-10UL
- ABCA_AB317594-100U
- Antigen Symbol:
- Tau Alzheimer’s Disease
- Tau Alzheimer’s Disease
- Antigen Name:
- MAPTL
- MAPTL
- Antigen Synonyms:
- MTBT1|TAU|MAPT|Microtubule-associated protein tau|Neurofibrillary tangle protein|Paired helical filament-tau|PHF-tau
- MTBT1|TAU|MAPT|Microtubule-associated protein tau|Neurofibrillary tangle protein|Paired helical filament-tau|PHF-tau
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 488
- Alexa Fluor® 488
- Clone:
- GT-38
- GT-38
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- Modification:
- phospho S396
- phospho S396
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 78928 Da
- 78928 Da
- Purification:
- Protein G affinity purification
- Protein G affinity purification
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle. Store in the dark.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle. Store in the dark.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-Fr
- IHC-Fr
Specifications
Frequently Bought Together