Anti-Gastric Mucin/MUC-6 Rabbit Monoclonal Antibody [clone: EPR20623]
: Abcam
New Product
Rabbit monoclonal [EPR20623] to Gastric Mucin/MUC-6 - BSA and Azide free.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Gastric Mucin/MUC-6
Clonality: Monoclonal
Clone: EPR20623
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB251578-100U
- ABCA_AB251578-1MG
- Antigen Symbol:
- Gastric Mucin/MUC-6
- Gastric Mucin/MUC-6
- Antigen Name:
- Mucin-6
- Mucin-6
- Antigen Synonyms:
- MUC-6|Gastric MUC6
- MUC-6|Gastric MUC6
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR20623
- EPR20623
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 257051 Da
- 257051 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Shipped at +4 °C. Store at +4 °C. Do Not Freeze.
- Shipped at +4 °C. Store at +4 °C. Do Not Freeze.
- Shipping Temperature:
- +4 °C
- +4 °C
- Tested Applications:
- IHC-P, mIHC
- IHC-P, mIHC
Frequently Bought Together