Anti-ERMN/Juxtanodin Rabbit Polyclonal Antibody
ABCA_AB243730-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:KIAA1189
- Antigen Symbol:ERMN/Juxtanodin
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:ERMN|Ermin|Juxtanodin|JN
- Molecular Weight:32783 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Recombinant Fragment Protein within Human ERMN aa 150 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Rabbit polyclonal to ERMN/Juxtanodin.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ERMN/Juxtanodin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human