Anti-CPS1 Rabbit Monoclonal Antibody [clone: EPR7493-3]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR7493-3] to CPS1 - BSA and Azide free.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CPS1
Clonality: Monoclonal
Clone: EPR7493-3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Specifications
- Catalog No:
- ABCA_AB248283-100U
- ABCA_AB248283-1MG
- Antigen Symbol:
- CPS1
- CPS1
- Antigen Name:
- Carbamoyl-phosphate synthetase I
- Carbamoyl-phosphate synthetase I
- Antigen Synonyms:
- CPSase I
- CPSase I
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR7493-3
- EPR7493-3
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 164939 Da
- 164939 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at +4 °C. Do not freeze.
- Store at +4 °C. Do not freeze.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Flow Cyt (Intra), IHC-P
- Flow Cyt (Intra), IHC-P
Specifications
Frequently Bought Together