Anti-COSMC Rabbit Polyclonal Antibody
ABCA_AB229831-50UL
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:HSPC067
- Antigen Symbol:COSMC
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:MSTP143|UNQ273/PRO310|C1GALT1C1|C1GALT1-specific chaperone 1|C38H2-like protein 1|Core 1 beta3-galactosyltransferase-specific molecular chaperone|C38H2-L1|C1Gal-T2|C1GalT2|Core 1 beta3-Gal-T2
- Molecular Weight:36382 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human C1GALT1C1 aa 1-200. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to COSMC.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: COSMC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human