Anti-Complement factor H-related protein 3 Rabbit Monoclonal Antibody [clone: EPR25759-77]
: Abcam
New Product
Rabbit monoclonal [EPR25759-77] to Complement factor H-related protein 3 - BSA and Azide free (Capture).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Complement factor H-related protein 3
Clonality: Monoclonal
Clone: EPR25759-77
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB305160-100U
- ABCA_AB305160-1MG
- Antigen Symbol:
- Complement factor H-related protein 3
- Complement factor H-related protein 3
- Antigen Name:
- CFHL3
- CFHL3
- Antigen Synonyms:
- FHR3|CFHR3|FHR-3|DOWN16|H factor-like protein 3
- FHR3|CFHR3|FHR-3|DOWN16|H factor-like protein 3
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR25759-77
- EPR25759-77
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 37323 Da
- 37323 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- sELISA
- sELISA
Frequently Bought Together