Anti-Importin 13 Rabbit Polyclonal Antibody
ABCA_AB95993-50UL
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:KIAA0724
- Antigen Symbol:Importin 13
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:RANBP13|IPO13|Importin-13|Imp13|Karyopherin-13|Ran-binding protein 13|Kap13
- Storage Buffer:Glycerol, tris, glycine
- Molecular Weight:108195 Da
- Storage Temperature:Upon delivery aliquot and store at −20 °C. Avoid repeated freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human IPO13 aa 900 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to Importin 13.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Importin 13
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human