Anti-Anterior Gradient 2 Rabbit Monoclonal Antibody [clone: EPR20164-278]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR20164-278] to Anterior Gradient 2 - BSA and Azide free.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Anterior Gradient 2
Clonality: Monoclonal
Clone: EPR20164-278
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Specifications
- Catalog No:
- ABCA_AB251500-100U
- ABCA_AB251500-1MG
- Antigen Symbol:
- Anterior Gradient 2
- Anterior Gradient 2
- Antigen Name:
- AG2
- AG2
- Antigen Synonyms:
- UNQ515/PRO1030|AGR2|Anterior gradient protein 2 homolog|AG-2|hAG-2|HPC8|Secreted cement gland protein XAG-2 homolog
- UNQ515/PRO1030|AGR2|Anterior gradient protein 2 homolog|AG-2|hAG-2|HPC8|Secreted cement gland protein XAG-2 homolog
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR20164-278
- EPR20164-278
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 19979 Da
- 19979 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at +4 °C. Do not freeze.
- Store at +4 °C. Do not freeze.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
Specifications
Frequently Bought Together