Anti-FRS2 Rabbit Monoclonal Antibody [clone: EPR14724]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR14724] to FRS2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: FRS2
Clonality: Monoclonal
Clone: EPR14724
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB183492-100U
- ABCA_AB183492-1ML
- Antigen Symbol:
- FRS2
- FRS2
- Antigen Name:
- Fibroblast growth factor receptor substrate 2
- Fibroblast growth factor receptor substrate 2
- Antigen Synonyms:
- FGFR substrate 2|FGFR-signaling adaptor SNT|Suc1-associated neurotrophic factor target 1|SNT-1
- FGFR substrate 2|FGFR-signaling adaptor SNT|Suc1-associated neurotrophic factor target 1|SNT-1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR14724
- EPR14724
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 57029 Da
- 57029 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Concentration:
- 0.112 mg/ml
- 0.112 mg/ml
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Flow Cyt (Intra)
- Flow Cyt (Intra)
Specifications
Frequently Bought Together