Order Entry
United States
ContactUsLinkComponent
Anti-MARCO Rabbit Monoclonal Antibody [clone: EPR26935-11]
Anti-MARCO Rabbit Monoclonal Antibody [clone: EPR26935-11]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-MARCO Rabbit Monoclonal Antibody [clone: EPR26935-11]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB314646-1ML
  • ABCA_AB314646-100U
  • ABCA_AB314646-20UL
  • Antigen Symbol:
  • MARCO
  • MARCO
  • MARCO
  • Antigen Name:
  • SCARA2
  • SCARA2
  • SCARA2
  • Antigen Synonyms:
  • Macrophage receptor MARCO|Macrophage receptor with collagenous structure|Scavenger receptor class A member 2
  • Macrophage receptor MARCO|Macrophage receptor with collagenous structure|Scavenger receptor class A member 2
  • Macrophage receptor MARCO|Macrophage receptor with collagenous structure|Scavenger receptor class A member 2
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR26935-11
  • EPR26935-11
  • EPR26935-11
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Molecular Weight:
  • 52658 Da
  • 52658 Da
  • 52658 Da
  • Purification:
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Storage Temperature:
  • Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
  • Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
  • Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • Shipped at +4 °C
  • Shipped at +4 °C
  • Shipped at +4 °C
  • Tested Applications:
  • ICC/IF, IHC-P, mIHC
  • ICC/IF, IHC-P, mIHC
  • ICC/IF, IHC-P, mIHC

Specifications

Frequently Bought Together