Anti-Junctional Adhesion Molecule 1/JAM-A Rabbit Monoclonal Antibody [clone: EPR23246-20]
: Abcam
New Product
Rabbit monoclonal [EPR23246-20] to Junctional Adhesion Molecule 1/JAM-A - BSA and Azide free (Capture).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Junctional Adhesion Molecule 1/JAM-A
Clonality: Monoclonal
Clone: EPR23246-20
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- ABCA_AB277253-100U
- ABCA_AB277253-1MG
- Antigen Symbol:
- Junctional Adhesion Molecule 1/JAM-A
- Junctional Adhesion Molecule 1/JAM-A
- Antigen Name:
- CD321
- CD321
- Antigen Synonyms:
- Jam1|Jcam|Jcam1|F11r|Junctional adhesion molecule A|JAM-A|Junctional adhesion molecule 1|JAM-1
- Jam1|Jcam|Jcam1|F11r|Junctional adhesion molecule A|JAM-A|Junctional adhesion molecule 1|JAM-1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR23246-20
- EPR23246-20
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 32424 Da
- 32424 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- sELISA
- sELISA
Frequently Bought Together