Order Entry
United States
ContactUsLinkComponent
 
Anti-Nogo A+Nogo D Rabbit Monoclonal Antibody [clone: EPR26286-56]
 :  Abcam
New Product
 
Anti-Nogo A+Nogo D Rabbit Monoclonal Antibody [clone: EPR26286-56]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCA_AB315792-100U
  • ABCA_AB315792-1ML
  • ABCA_AB315792-20UL
  • Antigen Symbol:
  • Nogo A+Nogo D
  • Nogo A+Nogo D
  • Nogo A+Nogo D
  • Antigen Name:
  • Kiaa0886
  • Kiaa0886
  • Kiaa0886
  • Antigen Synonyms:
  • Nogo|Reticulon-4|Neurite outgrowth inhibitor|Nogo protein
  • Nogo|Reticulon-4|Neurite outgrowth inhibitor|Nogo protein
  • Nogo|Reticulon-4|Neurite outgrowth inhibitor|Nogo protein
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR26286-56
  • EPR26286-56
  • EPR26286-56
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Molecular Weight:
  • 126613 Da
  • 126613 Da
  • 126613 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • 4 °C
  • Tested Applications:
  • Dot, Flow Cyt (Intra), ICC/IF, IHC-Fr, IHC-P
  • Dot, Flow Cyt (Intra), ICC/IF, IHC-Fr, IHC-P
  • Dot, Flow Cyt (Intra), ICC/IF, IHC-Fr, IHC-P

 

Frequently Bought Together