Anti-HLA-DR Mouse Monoclonal Antibody [clone: TAL 1B5]
: Abcam
New Product
Mouse monoclonal [TAL 1B5] to HLA-DR.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: TAL 1B5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
- Catalog No:
- ABCA_AB20181-100UG
- ABCA_AB20181-100UL
- ABCA_AB20181-1MG
- Antigen Symbol:
- HLA-DR
- HLA-DR
- HLA-DR
- Antigen Name:
- HLA-DRA1
- HLA-DRA1
- HLA-DRA1
- Antigen Synonyms:
- HLA-DRA|MHC class II antigen DRA
- HLA-DRA|MHC class II antigen DRA
- HLA-DRA|MHC class II antigen DRA
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- TAL 1B5
- TAL 1B5
- TAL 1B5
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 28621 Da
- 28621 Da
- 28621 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Storage Buffer:
- PBS
- PBS
- PBS
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- 4 °C
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P
- ICC/IF, IHC-P
Frequently Bought Together