Anti-LNK Rabbit Monoclonal Antibody [clone: EPR7342]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR7342] to LNK.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: LNK
Clonality: Monoclonal
Clone: EPR7342
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB154848-1ML
- ABCA_AB154848-100U
- Antigen Synonyms:
- SH2B adapter protein 3|Lymphocyte adapter protein|Lymphocyte-specific adapter protein Lnk|Signal transduction protein Lnk
- SH2B adapter protein 3|Lymphocyte adapter protein|Lymphocyte-specific adapter protein Lnk|Signal transduction protein Lnk
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Host:
- Rabbit
- Rabbit
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 63225 Da
- 63225 Da
- Purification:
- Tissue culture supernatant
- Tissue culture supernatant
- Storage Temperature:
- +4 °C
- +4 °C
- Shipping Temperature:
- Shipped at +4 °C. Store at –20ºC.
- Shipped at +4 °C. Store at –20ºC.
Specifications
Frequently Bought Together