Anti-Transferrin Receptor Mouse Monoclonal Antibody [clone: 2/Transferrin H1L1]
Supplier: Abcam
New Product
Certificates
About this item
Mouse monoclonal [2/Transferrin] to Transferrin Receptor.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Transferrin Receptor
Clonality: Monoclonal
Clone: 2/Transferrin H1L1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB300657-1MG
- ABCA_AB300657-100U
- ABCA_AB300657-10UG
- ABCA_AB300657-20UG
- Antigen Symbol:
- Transferrin Receptor
- Transferrin Receptor
- Transferrin Receptor
- Transferrin Receptor
- Antigen Name:
- CD71
- CD71
- CD71
- CD71
- Antigen Synonyms:
- Transferrin receptor protein 1|TR|TfR|TfR1|Trfr|T9|p90|TFRC
- Transferrin receptor protein 1|TR|TfR|TfR1|Trfr|T9|p90|TFRC
- Transferrin receptor protein 1|TR|TfR|TfR1|Trfr|T9|p90|TFRC
- Transferrin receptor protein 1|TR|TfR|TfR1|Trfr|T9|p90|TFRC
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 2/Transferrin H1L1
- 2/Transferrin H1L1
- 2/Transferrin H1L1
- 2/Transferrin H1L1
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 84871 Da
- 84871 Da
- 84871 Da
- 84871 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- ICC/IF
- ICC/IF
- ICC/IF
- ICC/IF
Specifications
Frequently Bought Together