Order Entry
United States
ContactUsLinkComponent
Anti-Histone H3 (acetyl K14) Rabbit Monoclonal Antibody [clone: EP964Y]
Anti-Histone H3 (acetyl K14) Rabbit Monoclonal Antibody [clone: EP964Y]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-Histone H3 (acetyl K14) Rabbit Monoclonal Antibody [clone: EP964Y]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB203952-1MG
  • ABCA_AB203952-100U
  • Antigen Symbol:
  • Histone H3
  • Histone H3
  • Antigen Name:
  • H3FA
  • H3FA
  • Antigen Synonyms:
  • HIST1H3A|H3C2|H3FL|HIST1H3B|H3C3|H3FC HIST1H3C|H3C4|H3FB|HIST1H3D|H3C6|H3FD|HIST1H3E|H3C7|H3FI|HIST1H3F|H3C8|H3FH|HIST1H3G|H3C10|H3FK|HIST1H3H|H3C11|H3FF|HIST1H3I|H3C12|H3FJ|HIST1H3J|HIST1H3C|H3FC|H3C1|Histone H3.1|Histone H3/a|Histone H3/b|Histone H3/c|Histone H3/d|Histone H3/f|Histone H3/h|Histone H3/i|Histone H3/j|Histone H3/k|Histone H3/l|H3K14ac
  • HIST1H3A|H3C2|H3FL|HIST1H3B|H3C3|H3FC HIST1H3C|H3C4|H3FB|HIST1H3D|H3C6|H3FD|HIST1H3E|H3C7|H3FI|HIST1H3F|H3C8|H3FH|HIST1H3G|H3C10|H3FK|HIST1H3H|H3C11|H3FF|HIST1H3I|H3C12|H3FJ|HIST1H3J|HIST1H3C|H3FC|H3C1|Histone H3.1|Histone H3/a|Histone H3/b|Histone H3/c|Histone H3/d|Histone H3/f|Histone H3/h|Histone H3/i|Histone H3/j|Histone H3/k|Histone H3/l|H3K14ac
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EP964Y
  • EP964Y
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • ChIP:
  • Yes
  • Yes
  • Modification:
  • acetyl K14
  • acetyl K14
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 15404 Da
  • 15404 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at 4 °C. Do not freeze.
  • Store at 4 °C. Do not freeze.
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • Tested Applications:
  • ICC/IF, IHC-P, PepArr
  • ICC/IF, IHC-P, PepArr

Specifications

Frequently Bought Together