Anti-Interferon alpha 2b Mouse Monoclonal Antibody [clone: 9D3]
Catalog # ABCA_AB9386-100UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:IFNA2A
- Antigen Symbol:Interferon alpha 2b
- Clonality:Monoclonal
- Clone:9D3
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 µl
- Neutralisation:Yes
- Form:Liquid
- Antigen Synonyms:IFNA2B|IFNA2C|IFNA2|Interferon alpha-2|IFN-alpha-2|Interferon alpha-A|LeIF A
- Storage Buffer:PBS
- Molecular Weight:21550 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C or –80 °C. Avoid freeze / thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human IFNA2.
- Tested Applications:sELISA
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [9D3] to Interferon alpha 2b.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Interferon alpha 2b
Clonality: Monoclonal
Clone: 9D3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human