Anti-Integrin alpha 2+beta 1 Mouse Monoclonal Antibody [clone: P1E6]
Catalog # ABCA_AB24697-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CD49b
- Antigen Symbol:Integrin alpha 2+beta 1
- Clonality:Monoclonal
- Clone:P1E6
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:ITGA2|Integrin alpha-2|CD49 antigen-like family member B|Collagen receptor|Platelet membrane glycoprotein Ia|VLA-2 subunit alpha|GPIa
- Storage Buffer:Ascites
- Molecular Weight:129295 Da
- Storage Temperature:Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped at +4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:Flow Cyt (Intra), ICC/IF
- Purification:Protein G Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [P1E6] to Integrin alpha 2+beta 1.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Integrin alpha 2+beta 1
Clonality: Monoclonal
Clone: P1E6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human