Anti-HIF-1 alpha (hydroxy P564) Rabbit Monoclonal Antibody [clone: EPR27307-108]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR27307-108] to HIF-1 alpha (hydroxyl P564) - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HIF-1 alpha
Clonality: Monoclonal
Clone: EPR27307-108
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB308638-1MG
- ABCA_AB308638-100U
- Antigen Symbol:
- HIF-1 alpha
- HIF-1 alpha
- Antigen Name:
- BHLHE78
- BHLHE78
- Antigen Synonyms:
- MOP1|PASD8|HIF1A|Hypoxia-inducible factor 1-alpha|HIF-1-alpha|HIF1-alpha|ARNT-interacting protein|Basic-helix-loop-helix-PAS protein MOP1|Class E basic helix-loop-helix protein 78|Member of PAS protein 1|PAS domain-containing protein 8
- MOP1|PASD8|HIF1A|Hypoxia-inducible factor 1-alpha|HIF-1-alpha|HIF1-alpha|ARNT-interacting protein|Basic-helix-loop-helix-PAS protein MOP1|Class E basic helix-loop-helix protein 78|Member of PAS protein 1|PAS domain-containing protein 8
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR27307-108
- EPR27307-108
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Modification:
- hydroxy P564
- hydroxy P564
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 92670 Da
- 92670 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Dot, Flow Cyt (Intra), ICC/IF
- Dot, Flow Cyt (Intra), ICC/IF
Specifications
Frequently Bought Together