Order Entry
United States
ContactUsLinkComponent
Anti-CSF-1-R Rat Monoclonal Antibody [clone: 12-3A3-1B10] (Biotin)
Anti-CSF-1-R Rat Monoclonal Antibody [clone: 12-3A3-1B10] (Biotin)
Catalog # ABCA_AB95730-50UG
Supplier:  Abcam
New Product
undefined
Anti-CSF-1-R Rat Monoclonal Antibody [clone: 12-3A3-1B10] (Biotin)
Catalog # ABCA_AB95730-50UG
Supplier:  Abcam
Supplier Number:  AB95730-50UG
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD115
  • Antigen Symbol:
    CSF-1-R
  • Clonality:
    Monoclonal
  • Clone:
    12-3A3-1B10
  • Conjugation:
    Biotin
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    50 µg
  • Form:
    Liquid
  • Antigen Synonyms:
    FMS|CSF1R|Macrophage colony-stimulating factor 1 receptor|CSF-1 receptor|Proto-oncogene c-Fms|CSF-1R|M-CSF-R
  • Storage Buffer:
    PBS, sodium chloride
  • Molecular Weight:
    107984 Da
  • Storage Temperature:
    Store at 4 °C short term (1 - 2 weeks). Store at −20 or −80 °C. Avoid freeze/thaw cycle.
  • Concentration:
    0.5 mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Virus preparation containing CSF1R protein. The exact immunogen used to generate this antibody is proprietary information.
  • Purification:
    Protein G affinity purification
  • Packaging:
    Vial

Specifications

About this item

Biotin Rat monoclonal [12-3A3-1B10] to CSF-1-R.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CSF-1-R
Clonality: Monoclonal
Clone: 12-3A3-1B10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human