Anti-alpha 1 Sodium Potassium ATPase Mouse Monoclonal Antibody [clone: 464.6]
Catalog # ABCA_AB7671-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Sodium/potassium-transporting ATPase subunit alpha-1
- Antigen Symbol:alpha 1 Sodium Potassium ATPase
- Clonality:Monoclonal
- Clone:464.6
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Mouse,Rat,Rabbit,Human,Pig
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Na(+)/K(+) ATPase alpha-1 subunit|Sodium pump subunit alpha-1|ATP1A1
- Storage Buffer:PBS
- Molecular Weight:112896 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested Applications:ICC/IF, IHC-P
- Purification:Protein G affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [464.6] to alpha 1 Sodium Potassium ATPase.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: alpha 1 Sodium Potassium ATPase
Clonality: Monoclonal
Clone: 464.6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat;Rabbit;Human;Pig