Anti-Hsp60 Mouse Monoclonal Antibody [clone: LK-1]
Catalog # ABCA_AB59457-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:HSPD1
- Antigen Symbol:Hsp60
- Clonality:Monoclonal
- Clone:LK-1
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Mouse,Rat,Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:60 kDa chaperonin|Chaperonin 60|Heat shock protein 60|Heat shock protein family D member 1|HuCHA60|Mitochondrial matrix protein P1|P60 lymphocyte protein|CPN60|HSP-60
- Storage Buffer:PBS, glycerol
- Molecular Weight:61055 Da
- Storage Temperature:Store at −20 °C. Stable for 12 months at −20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human HSPD1
- Tested Applications:ICC/IF, IHC-P
- Purification:Protein G affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [LK-1] to Hsp60.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Hsp60
Clonality: Monoclonal
Clone: LK-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat;Human