Anti-DDX53/CT26 Rabbit Monoclonal Antibody [clone: EPR27384-107]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR27384-107] to DDX53/CT26 - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: DDX53/CT26
Clonality: Monoclonal
Clone: EPR27384-107
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB313334-1MG
- ABCA_AB313334-100U
- Antigen Symbol:
- DDX53/CT26
- DDX53/CT26
- Antigen Name:
- CAGE
- CAGE
- Antigen Synonyms:
- DDX53|Probable ATP-dependent RNA helicase DDX53|Cancer-associated gene protein|Cancer/testis antigen 26|DEAD box protein 53|DEAD box protein CT26
- DDX53|Probable ATP-dependent RNA helicase DDX53|Cancer-associated gene protein|Cancer/testis antigen 26|DEAD box protein 53|DEAD box protein CT26
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR27384-107
- EPR27384-107
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 71154 Da
- 71154 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together