Anti-THSD7A Mouse Monoclonal Antibody [clone: CL3778]
Catalog # ABCA_AB243031-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:KIAA0960
- Antigen Symbol:THSD7A
- Clonality:Monoclonal
- Clone:CL3778
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Thrombospondin type-1 domain-containing protein 7A
- Molecular Weight:185363 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipping Temperature:+4 °C
- Immunogen:Recombinant Fragment Protein within Human THSD7A aa 200-400. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [CL3778] to THSD7A.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: THSD7A
Clonality: Monoclonal
Clone: CL3778
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human