Anti-NKG2D Rabbit Recombinant Multiclonal Antibody [clone: RM1173]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit recombinant multiclonal [RM1173] to NKG2D.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: NKG2D
Clonality: Recombinant Multiclonal
Clone: RM1173
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB319162-100U
- ABCA_AB319162-40UL
- ABCA_AB319162-1ML
- Antigen Symbol:
- NKG2D
- NKG2D
- NKG2D
- Antigen Name:
- CD314
- CD314
- CD314
- Antigen Synonyms:
- D12S2489E|KLRK1|NKG2-D type II integral membrane protein|Killer cell lectin-like receptor subfamily K member 1|NK cell receptor D|NKG2-D-activating NK receptor
- D12S2489E|KLRK1|NKG2-D type II integral membrane protein|Killer cell lectin-like receptor subfamily K member 1|NK cell receptor D|NKG2-D-activating NK receptor
- D12S2489E|KLRK1|NKG2-D type II integral membrane protein|Killer cell lectin-like receptor subfamily K member 1|NK cell receptor D|NKG2-D-activating NK receptor
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Recombinant Multiclonal
- Recombinant Multiclonal
- Recombinant Multiclonal
- Clone:
- RM1173
- RM1173
- RM1173
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Liquid
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P
- IHC-P
Specifications
Frequently Bought Together