Order Entry
United States
ContactUsLinkComponent
Anti-Islet 1 Mouse Monoclonal Antibody [clone: 1B1]
Anti-Islet 1 Mouse Monoclonal Antibody [clone: 1B1]
Catalog # ABCA_AB86501-100UL
Supplier:  Abcam
New Product
Anti-Islet 1 Mouse Monoclonal Antibody [clone: 1B1]
Catalog # ABCA_AB86501-100UL
Supplier:  Abcam
Supplier Number:  AB86501-100UL
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Insulin gene enhancer protein ISL-1
  • Antigen Symbol:
    Islet 1
  • Clonality:
    Monoclonal
  • Clone:
    1B1
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Western Blot:
    Yes
  • Size:
    100 µl
  • Form:
    Liquid
  • Antigen Synonyms:
    Islet-1|ISL1
  • Storage Buffer:
    Ascites
  • Molecular Weight:
    39036 Da
  • Storage Temperature:
    Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping Temperature:
    Shipped at +4 °C
  • Immunogen:
    Recombinant Fragment Protein within Human ISL1 aa 150 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
  • Tested Applications:
    ICC/IF, IHC-P
  • Purification:
    Protein G Affinity purification
  • Packaging:
    Vial

Specifications

About this item

Mouse monoclonal [1B1] to Islet 1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Islet 1
Clonality: Monoclonal
Clone: 1B1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human