Anti-GMNN Mouse Monoclonal Antibody [clone: 1A8]
Catalog # ABCA_AB104306-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Geminin
- Antigen Symbol:GMNN
- Clonality:Monoclonal
- Clone:1A8
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Storage Buffer:PBS
- Molecular Weight:23565 Da
- Storage Temperature:Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human GMNN aa 100 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Tissue culture supernatant
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [1A8] to Geminin.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: GMNN
Clonality: Monoclonal
Clone: 1A8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human