Anti-SERPINB14 Rabbit Monoclonal Antibody [clone: EPR27117-90]
: Abcam
New Product
Rabbit monoclonal [EPR27117-90] to Ovalbumin.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SERPINB14
Clonality: Monoclonal
Clone: EPR27117-90
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
- Catalog No:
- ABCA_AB306591-100U
- ABCA_AB306591-1ML
- ABCA_AB306591-20UL
- Antigen Symbol:
- SERPINB14
- SERPINB14
- SERPINB14
- Antigen Name:
- Ovalbumin
- Ovalbumin
- Ovalbumin
- Antigen Synonyms:
- Allergen Gal d II|Egg albumin|Plakalbumin
- Allergen Gal d II|Egg albumin|Plakalbumin
- Allergen Gal d II|Egg albumin|Plakalbumin
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- EPR27117-90
- EPR27117-90
- EPR27117-90
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 42881 Da
- 42881 Da
- 42881 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- 4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF
- Flow Cyt (Intra), ICC/IF
- Flow Cyt (Intra), ICC/IF
Frequently Bought Together